Anti-GAL4 Rabbit Polyclonal Antibody

Supplier: Proteintech
13391-1-AP-150UL
10087-340EA 402 USD
10087-340
Anti-GAL4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A; IF:COLO 320 Cells, 1:10-1:100

Type: Primary
Antigen: Galectin 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR