Anti-HSPH1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13383-1-AP-150UL
10087-638EA 402 USD
10087-638
Anti-HSPH1 Rabbit Polyclonal Antibody
Antibodies
HSP105, also known as HSP110 or HSPH1, belongs to the heat shock protein (HSP) family. Human HSP105 is a high-molecular-weight chaperone protein expressed at constitutively low levels as a cytoplasmic α-isoform and as an inducible nuclear β-isoform on exposure to various forms of stress. HSP105 is constitutively overexpressed in several solid tumors, including melanoma, breast, thyroid, and gastroenteric cancers, and exerts antiapoptotic functions. Recently HSP105 has been identified as a novel candidate biomarker of lymphoma aggressiveness. This antibody recognizes both HSP105α and HSP105β isoforms. Western blot analysis using this antibody detected a major band around 100-110kD in Jurkat cells.

Western Blot: MCF7 Cells, 1:500-1:5000; IF: Hela Cells, 1:20-1:200; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: HSPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR