Anti-GLRX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13381-1-AP-150UL
10087-248EA 402 USD
10087-248
Anti-GLRX2 Rabbit Polyclonal Antibody
Antibodies
GLRX2(Glutaredoxin-2) is also named as GRX2 and belongs to the glutaredoxin family. It facilitates the maintenance of cellular redox homeostasis upon treatment with apoptotic agents, thereby preventing cardiolipin oxidation and cytochrome C release. The GLRX2, which exists in a dynamic equilibrium of enzymatically active monomers and quiescent dimers,is a 16-kDa protein directed either to the nucleus or to the mitochondria through differential splicing of the first exon. It has 2 isoforms produced by alternative splicing.

Western Blot:A549 Cells, 1:200-1:1000

Type: Primary
Antigen: GLRX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR