Anti-IFIT5 Rabbit Polyclonal Antibody

Supplier: Proteintech
13378-1-AP-150UL
10088-600EA 402 USD
10088-600
Anti-IFIT5 Rabbit Polyclonal Antibody
Antibodies
IFIT5, also named as Interferon-induced 58 kDa protein or RI58, is a 482 amino acid protein, which contains eight TPR repeats and belongs to the IFIT family. IFIT5 localizes in the Cell projection. Interferon-induced RNA-binding protein that specifically binds single-stranded RNA bearing a 5'-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs.

Western Blot:A375 Cells, 1:200-1:2000; IHC: Human skin cancer Tissue, 1:20-1:200

Type: Primary
Antigen: IFIT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR