Anti-TBRG1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13346-1-AP-150UL
10095-656EA 402 USD
10095-656
Anti-TBRG1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:BxPC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: TBRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR