Anti-TAF1C Rabbit Polyclonal Antibody

Supplier: Proteintech
13341-1-AP-150UL
10095-408EA 402 USD
10095-408
Anti-TAF1C Rabbit Polyclonal Antibody
Antibodies
Western Blot: HT-1080 Cells, 1:200-1:2000

Type: Primary
Antigen: TAF1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR