Anti-LENG8 Rabbit Polyclonal Antibody

Supplier: Proteintech
13336-1-AP-150UL
10089-594EA 402 USD
10089-594
Anti-LENG8 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A2780 Cells, 1:500-1:5000

Type: Primary
Antigen: LENG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR