Anti-P-selectin Rabbit Polyclonal Antibody

Supplier: Proteintech
13304-1-AP-150UL
10094-284EA 402 USD
10094-284
Anti-P-selectin Rabbit Polyclonal Antibody
Antibodies
P-selectin, also known as GMP140 or CD62P, is a transmembrane glycoprotein that mediates the interaction of activated endothelial cells or platelets with leukocytes. It is an adhesion molecule involved in the pathogenesis of inflammation, thrombosis, and oncogenesis. P-selectin is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. Upon cell activation by agonists, P-selectin is transported rapidly to the cell surface.

IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: P-selectin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR