Anti-FGFBP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13254-1-AP-150UL
10086-978EA 402 USD
10086-978
Anti-FGFBP2 Rabbit Polyclonal Antibody
Antibodies
Fibroblast growth factor-binding protein 2 (FGFBP2), also known as 37 kDa killer-specific secretory protein (KSP37), is a member of the fibroblast growth factor binding protein family. FGFBP2 was identified as a Th1/Tc1 specific secretory protein is expressed preferentially in cytotoxic T lymphocytes (CTL) and natural killer (NK) cells and might be involved in essential processes of CTL-mediated immunity. An increase expression of FGFBP2 may be associated with atopic asthma and mild extrinsic asthma. Recently, an observation revealed that FGFBP2 correlate strongly with histology, stage and outcome in ovarian, and it might be a potential prognostic biomarker. The observed MW of FGFBP2 (37 kDa) varies from the predicated MW of 28 kDa, which may be due to the specific space conformation caused by internal disulfide bonds.

Western Blot:SH-SY5Y Cells, 1:1000-1:10000; IF:BxPC-3 Cells, 1:10-1:100

Type: Primary
Antigen: FGFBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR