Anti-CYP4F12 Rabbit Polyclonal Antibody

Supplier: Proteintech
13243-1-AP-150UL
10085-542EA 402 USD
10085-542
Anti-CYP4F12 Rabbit Polyclonal Antibody
Antibodies
CYP4F12(Cytochrome P450 4F12) Catalyzes leukotriene B4 omega-hydroxylation and arachidonic acid omega-hydroxylation but with an activity much lower than that of CYP4F2.It expresses a band of 60KDa in adult and foetal liver, kidney, placenta at term, seminal vesicles, the prostate gland.And it can be phosphorylated in vivo.

Western Blot: Human Placenta Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CYP4F12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR