Anti-UBA6 Rabbit Polyclonal Antibody

Supplier: Proteintech
13211-1-AP-150UL
10096-558EA 402 USD
10096-558
Anti-UBA6 Rabbit Polyclonal Antibody
Antibodies
UBA6(Ubiquitin-like modifier-activating enzyme 6) is also named as MOP4, UBE1L2 and belongs to the ubiquitin-activating E1 family. It probably acts as a single molecule like UBE1 and not as a heterodimeric E1 complex, as formed by AOS1/Uba2 and APP-BP1/Uba3. UBA6 is even more restricted in distribution because it is only found in vertebrates but not in lower organisms so that it might have a specialized role in tissues of higher organisms, and in particular, in the testis, where it is expressed by far most abundantly.. It has 4 isoforms produced by alternative splicing. This antibody recognizes the isoform 1(118 kda) and isoform 2(66 kDa) of UBA6.

Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: UBA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR