Anti-PPP3R1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13210-1-AP-150UL
10092-726EA 402 USD
10092-726
Anti-PPP3R1 Rabbit Polyclonal Antibody
Antibodies
Protein phosphatase 3 (PPP3, formerly PP2B, Calcineurin), a serine/ threonine protein phosphatase, is a heterodimer composed of one catalytic subunit (PPP3C, Calcineurin A) and one regulatory subunit (PPP3R, Calcineurin B). PPP3R has two isoforms, PPP3R1 and PPP3R2. PPP3Rl is ubiquitously expressed in different tissues and is coded by the PPP3R1 gene which was mapped recently to human chromosome 2. PPP3Rl has four functional calcium-binding sites.

Western Blot:K-562 Cells, 1:200-1:1000; IHC: Human gliomas Tissue, 1:20-1:200; IP: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: PPP3R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR