Anti-DCLRE1B Rabbit Polyclonal Antibody

Supplier: Proteintech
13203-1-AP-150UL
10085-700EA 402 USD
10085-700
Anti-DCLRE1B Rabbit Polyclonal Antibody
Antibodies
Interstrand cross-links (ICLs) is an extremely cytotoxic form of DNA damage that covalently tethers the two strands of the double helix, which inhibits fundamental nuclear functions, including transcription and DNA replication . DCLRE1B is one of several evolutionarily conserved genes involved in repair of interstrand cross-links . Furthermore, DCLRE1B was identified as a TRF2-interacting protein and, has a role in telomere maintenance in association with the Shelterin complex, preventing non-homologous end-joining-mediated chromosome fusions protecting leading strand telomeres .

Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: DCLRE1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR