Anti-AXL Rabbit Polyclonal Antibody

Supplier: Proteintech
13196-1-AP-150UL
10083-426EA 402 USD
10083-426
Anti-AXL Rabbit Polyclonal Antibody
Antibodies
Axl, also known as Ufo or Ark, is a receptor tyrosine kinase of the TAM (Tyro3, Axl and Mer) family. Axl is a single-pass type I membrane protein containing two fibronectin type-III domains, two Ig-like C2-type domains and one protein kinase domain. Axl mRNA is widely expressed with highest levels in the heart and skeletal muscle. Gas6 has been identified as a ligand for human Axl. Gas6/Axl signaling plays a role in cell survival, cell proliferation, migration and differentiation. This antibody detects a single band at 140 kDa, which is considered to represent the auto-phosphorylated form of endogenous Axl .

  • Western Blot: HepG2 Cells, 1:500-1:5000; IP: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: AXL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR