Anti-FXR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13194-1-AP-150UL
10085-966EA 402 USD
10085-966
Anti-FXR1 Rabbit Polyclonal Antibody
Antibodies
Tumour necrosis factor-alpha (TNF-alpha) is a key mediator of inflammation in host defence against infection and in autoimmune disease. Its production is controlled post-transcriptionally by multiple RNA-binding proteins that interact with the TNF-alpha AU-rich element and regulate its expression; Fragile X mental retardation-related protein 1 (FXR1) is one of these. FXR1 (fragile-X-mental retardation-related protein 1) are RNA-binding proteins that have been demonstrated to impact miRNA-mediated, post-transcriptional gene regulation, and required for embryonic and postnatal development of muscle tissue. It can regulate intracellular transport and local translation of certain mRNAs

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IP: Mouse Skeletal Muscle Tissue, 1:200-1:2000; IF:Ethacrynic acid treated HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: FXR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR