Anti-PSMA/GCPII Rabbit Polyclonal Antibody

Supplier: Proteintech
13163-1-AP-150UL
10092-886EA 402 USD
10092-886
Anti-PSMA/GCPII Rabbit Polyclonal Antibody
Antibodies
PSMA(Prostate-specific membrane antigen) is also named as FOLH1, FOLH, NAALAD1, PSM and belongs to the peptidase M28 family. PSMA is a 100-120 kDa integral transmembrane glycoprotein, considered to be a highly specific marker of the prostate gland, and has successfully been used as a marker of circulating prostatic epithelial cells. It is involved in conversion of the major neurotransmitter (NAAG) to NAA and free glutamate. It has 8 isoforms produced by alternative splicing.

Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PSMA/GCPII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR