Anti-FBXW11 Rabbit Polyclonal Antibody

Supplier: Proteintech
13149-1-AP-150UL
10086-422EA 402 USD
10086-422
Anti-FBXW11 Rabbit Polyclonal Antibody
Antibodies
FBXW11(F-box/WD repeat-containing protein 11) is also named as BTRCP2, FBW1B, FBXW1B, KIAA0696,HOS.It mediates the ubiquitination of phosphorylated NFKBIA, which degradation frees the associated NFKB1 to translocate into the nucleus and to activate transcription.It has 3 isoforms produced by alternative splicing.

Western Blot:SGC-7901 Cells, 1:1000-1:10000

Type: Primary
Antigen: FBXW11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR