Anti-FOXM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13147-1-AP-150UL
10087-120EA 537.7 USD
10087-120
Anti-FOXM1 Rabbit Polyclonal Antibody
Antibodies
Forkhead box M1 (FOXM1), also named HNF-3, HFH-11 or Trident, belongs to a superfamily of Fox transcription factors. FOXM1 is strongly associated with cell proliferation, it plays a important role in cell cycle progression by regulating both G1/S and G2/M phases, it is also involved in DNA damage repair, angiogenesis, apoptosis and tissue regeneration. Recently, crucial roles of this transcription factor were found in the development and progression of many cancers including colorectal, lung, prostate, liver and breast cancer. FOXM1 levels are significantly higher and associated with tumor grade in most human tumors, rendering it a potential target of cancer diagnosis and therapies. Catalog#13147-1-AP is rabbit polyclonal antibody raised against C-terminal of human FOXM1. And this antibody can recognize three kinds of FOXM1,FOXM1-pp(~95-110kDa), FOXM1-B(isoform2,~83kDa) and FOXM1A(isoform4, ~90kDa)

Western Blot: Mouse Thymus Tissue, 1:500-1:5000; IP:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: FOXM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR