Anti-TEAD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
13120-1-AP-150UL
10095-766EA 402 USD
10095-766
Anti-TEAD3 Rabbit Polyclonal Antibody
Antibodies
TEAD3, others named ranscriptional enhancer factor 5(TEF5), a member of family of transcription factors that plays a key role in the Hippo signaling pathway, suggesting involve in organ and tumor suppression by restricting proliferation and promoting. TEAD3 contains the TEA/ATTS DNA-binding domain. Human TEAD3 has a strong mRNA expression in placenta and choriocarcinoma cells(JEG-3), but low in Hela, HepG2 and MCF-7. This is a rabbit polyclonal anti-TEAD3 antibody raised against part of C-terminal chain of human TEAD3.

Western Blot: Human Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: TEAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR