Anti-PYCR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13108-1-AP-150UL
10081-420EA 459.7 USD
10081-420
Anti-PYCR1 Rabbit Polyclonal Antibody
Antibodies
PYCR1,also named as P5CR1, belongs to the pyrroline-5-carboxylate reductase family. It is a housekeeping enzyme that catalyzes the last step in proline biosynthesis. PYCR1 can utilize both NAD and NADP, but has higher affinity for NAD. It is involved in the cellular response to oxidative stress. Mutation in PYCR1 will cause ARCL type II(ARCL2B ). Some mutation will cause DeBarsy syndrome (DBS) which is characterized by progeroid features, ophthalmological abnormalities, intrauterine growth retardation, and cutis laxa. The MW of PYCR1 is about 33-35kd.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: PYCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR