Anti-DNAJC10 Rabbit Polyclonal Antibody

Supplier: Proteintech
13101-1-AP-150UL
10085-996EA 402 USD
10085-996
Anti-DNAJC10 Rabbit Polyclonal Antibody
Antibodies
DNAJC10(DnaJ homolog subfamily C member 10)is also named as ERDJ5. The human ER-resident protein (ERdj5) ubiquitously expresses and is abundant in secretory tissues such as pancreas and testis and it may be involved in assisting protein folding and quality control in the ER. The deduced 793-amino acid protein has a calculated molecular mass of about 91 kD. ERDJ5 contains an N-terminal hydrophobic sequence, followed by a type III DnaJ domain, 4 thioredoxin-like domains, and a C-terminal KDEL endoplasmic reticulum (ER) retention signal. It can be modified by N-linked glycosylation. This antibody is specific to DNAJC10.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human pancreas cancer, 1:20-1:200

Type: Primary
Antigen: DNAJC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR