Anti-IFNAR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13083-1-AP-150UL
10088-608EA 402 USD
10088-608
Anti-IFNAR1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:500-1:5000

Type: Primary
Antigen: IFNAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR