Anti-CDK2AP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13060-2-AP-150UL
10084-518EA 402 USD
10084-518
Anti-CDK2AP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: CDK2AP1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR