Anti-G3BP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13057-2-AP-150UL
10087-216EA 537.7 USD
10087-216
Anti-G3BP1 Rabbit Polyclonal Antibody
Antibodies
GAP SH3 Binding Protein 1 (G3BP1), also named as G3BP, is an effector of stress granule (SG) assembly. SG biology plays an important role in the pathophysiology of TDP-43 in ALS and FTLD-U. G3BP1 can be used as a marker of SG. It has been shown to function downstream of Ras and play a role in RNA metabolism, signal transduction, and proliferation. G3BP1 is a ubiquitously expressed protein that localizes to the cytoplasm in proliferating cells and to the nucleus in non-proliferating cells. G3BP1 has recently been implicated in cancer biology.

Western Blot:A431 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HEK-293 Cells, 1:500-1:5000; FC: HeLa Cells, N/A

Type: Primary
Antigen: G3BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR