Anti-CLDN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13050-1-AP-150UL
10084-822EA 402 USD
10084-822
Anti-CLDN1 Rabbit Polyclonal Antibody
Antibodies
Claudins are a family of proteins that are the most important components of the tight junctions, where they establish the paracellular barrier that controls the flow of molecules in the intercellular space between the cells of an epithelium. 23 claudins have been identified. They are small (20-27 kilodalton (kDa)) proteins with very similar structure. They have four transmembrane domains, with the N-terminus and the C-terminus in the cytoplasm. Claudin-1 is expressed primarily in keratinocytes and normal mammary epithelial cells.

Western Blot: Mouse Thymus Tissue, 1:200-1:2000; IHC: Human skin Tissue, 1:20-1:200

Type: Primary
Antigen: CLDN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR