Anti-ELAVL4 Rabbit Polyclonal Antibody

Supplier: Proteintech
13032-1-AP-150UL
10086-466EA 402 USD
10086-466
Anti-ELAVL4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human cerebellum Tissue, 1:1000-1:10000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: ELAVL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR