Anti-FBX15 Rabbit Polyclonal Antibody

Supplier: Proteintech
13024-1-AP-150UL
10086-896EA 402 USD
10086-896
Anti-FBX15 Rabbit Polyclonal Antibody
Antibodies
F-box proteins have been shown to be critical for the ubiquitin-mediated degradation of cellular regulatory proteins, and they are a family of eukaryotic proteins characterized by an approximately 40 amino acid motif. SCF complex, a class of ubiquitin ligases, consists of invariable components, Skp1 and Cullin, and variable components of F-box proteins, which have a primary role in determining substrate specificity. The F-box proteins are divided into 3 classes: FBWs containing WD-40 domains, FBLs containing leucine-rich repeats, and FBXs containing either different protein-protein interaction modules or no recognizable motifs. FBOX15 (or FBX15) belongs to the FBXs class, and 2 isoforms produced by alternative splicing has been described. This antibody is expected detect both isoforms.

Western Blot:A2780 Cells, 1:500-1:5000

Type: Primary
Antigen: FBX15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR