Anti-CRTAC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
13001-1-AP-150UL
10085-210EA 402 USD
10085-210
Anti-CRTAC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: CRTAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR