Anti-NF-L Rabbit Polyclonal Antibody

Supplier: Proteintech
12998-1-AP-150UL
10090-646EA 524.48 USD
10090-646
Anti-NF-L Rabbit Polyclonal Antibody
Antibodies
NEFL, also named as NF68 and NFL, belongs to the intermediate filament family. Neurofilaments are the 10nm intermediate filaments found specifically in neurons. They are a major component of the cell's cytoskeleton, and provide support for normal axonal radial growth. Neurofilaments usually contain three intermediate filament proteins: L, M, and H which are involved in the maintenance of neuronal caliber. The names given to the three major neurofilament subunits are based upon the apparent molecular weight of the mammalian subunits on SDS-PAGE:NF-L, 65-68kd; NF-M,145-160kd and NF-H, 200-220kd.This antibody is specific to NEFL. It has a very weak reaction to NEFM.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IHC: Human Brain Tissue, 1:20-1:200; IF:NIH/3T3 Cells, 1:10-1:100

Type: Primary
Antigen: NF-L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR