Anti-NEU4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12995-1-AP-150UL
10091-046EA 402 USD
10091-046
Anti-NEU4 Rabbit Polyclonal Antibody
Antibodies
NEU4, also named as LP5125, belongs to a family of glycohydrolytic enzymes, which remove terminal sialic acid residues from various sialo derivatives, such as glycoproteins, glycolipids, oligosaccharides, and gangliosides. It may function in lysosomal catabolism of sialylated glycoconjugates. NEU4 has a broad substrate specificity being active on glycoproteins, oligosaccharides and sialylated glycolipids.

IHC: Human Liver Tissue, 1:20-1:200; Western Blot:L02 Cells, 1:500-1:5000

Type: Primary
Antigen: NEU4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR