Anti-CLASP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12942-1-AP-150UL
10084-820EA 402 USD
10084-820
Anti-CLASP2 Rabbit Polyclonal Antibody
Antibodies
CLASP2 (CLIP-associating protein 2) is a brain specific protein belongs to CLASP family. CLASP1 and CLASP2 regulate microtubule plus-end dynamics in brine cortex. CLASP2 is required for the orientation of stabilized microtubules toward the leading edge, It may act at the cell cortex to enhance the frequency of rescue of depolymerizing microtubules by attaching their plus-ends to cortical platforms. Phosphorylation of CLASP2 by GSK3B may negatively regulate binding to microtubule lattices in lamella.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: CLASP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR