Anti-PSME2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12937-2-AP-150UL
10092-990EA 402 USD
10092-990
Anti-PSME2 Rabbit Polyclonal Antibody
Antibodies
The principal function of the proteasome is targeted degradation of intracellular proteins. Activity of the 20S proteasome is controlled by regulatory complexes that bind to the ends of the cylindrical proteasome. 11S regulator (REG or PA28), is a complex of 28 kDa subunits that is thought to activate proteasomes toward the production of antigenic peptides. Human PSME1 and PSME2 genes encode the two proteasome activators PA28 alpha and beta, respectively, which have been implicated in antigen processing for loading class I MHC molecules. The PA28 activator complex enhances the generation of class I binding peptides by altering the cleavage pattern of the proteasome.

Western Blot: HepG2 Cells, 1:500-1:5000; IP: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: PSME2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR