Anti-SKAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12926-1-AP-150UL
10094-664EA 402 USD
10094-664
Anti-SKAP2 Rabbit Polyclonal Antibody
Antibodies
SKAP2 (src kinase associated phosphoprotein 2), also named PRAP, contains PH domain and SH3 domain. It is localized in cytoplasm and was expressed in ubiquitously in various tissues including lung, skeletal muscle, and spleen, but not detectable in thymus and T cell lymphoma cells. SKAP2 may negatively regulate alpha-synuclein phosphorylation following cell stress. SKAP2 may be involved in B-cell and macrophage adhesion processes by coupling the B-cell receptor (BCR) to integrin activation.

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IP: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: SKAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR