Anti-TGM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12912-3-AP-150UL
10095-868EA 537.7 USD
10095-868
Anti-TGM1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Thymus Tissue, 1:200-1:2000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: TGM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR