Anti-CYP2W1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12904-1-AP-150UL
10085-530EA 402 USD
10085-530
Anti-CYP2W1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: CYP2W1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR