Anti-SH2D3C Rabbit Polyclonal Antibody

Supplier: Proteintech
12882-1-AP-150UL
10094-522EA 402 USD
10094-522
Anti-SH2D3C Rabbit Polyclonal Antibody
Antibodies
SH2 domain-containing protein 3C (SH2D3C, or NSP3) gene encodes an adaptor protein and member of a cytoplasmic protein family involved in cell migration. The encoded protein was identified as an Eph receptor-binding protein which may be a positive regulator of TCR signaling . SH2D3C is an adaptor protein that has been implicated in T cell activation and shown to associate with different components of the integrin signaling pathway ex vivo, moreover, its primary in vivo role of Sh2d3c in regulating B cell development and function has also been demonstrated .

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: SH2D3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR