Anti-LILRB5 Rabbit Polyclonal Antibody

Supplier: Proteintech
12881-1-AP-150UL
10089-416EA 402 USD
10089-416
Anti-LILRB5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:1000-1:10000

Type: Primary
Antigen: LILRB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR