Anti-UGCG Rabbit Polyclonal Antibody

Supplier: Proteintech
12869-1-AP-150UL
10096-730EA 402 USD
10096-730
Anti-UGCG Rabbit Polyclonal Antibody
Antibodies
UGCG, also named as GCS and GLCT-1, belongs to the glycosyltransferase 2 family. It catalyzes the first glycosylation step in glycosphingolipid biosynthesis, the transfer of glucose to ceramide. UGCG may also serve as a "flippase". It is up-regulated at the transcriptional level during keratinocyte differentiation. UGCG is a membrane protein. The calculated MW is 45kd. In SDS-PAGE, it is about 50-55kd.

Western Blot:A375 Cells, 1:500-1:5000; IHC: Human Lymphoma, 1:20-1:200

Type: Primary
Antigen: UGCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR