Anti-GP1BA Rabbit Polyclonal Antibody

Supplier: Proteintech
12860-1-AP-150UL
10084-352EA 524.48 USD
10084-352
Anti-GP1BA Rabbit Polyclonal Antibody
Antibodies
GP1BA, also named as CD42b , is a surface membrane protein of platelets, participates in the formation of platelet plugs by binding to the A1 domain of vWF, which is already bound to the subendothelium. It has a heterodimer form which composed of GP1BA and GP1BB. It is 145kd in platelet. It is Glycocalicin and phopho and showed 80-85kd in Jurkat.

Western Blot:A431 Cells, 1:200-1:2000; IHC: Human spleen Tissue, 1:20-1:200; IP:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: CD42b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR