Anti-PHYH Rabbit Polyclonal Antibody

Supplier: Proteintech
12858-1-AP-150UL
10092-214EA 402 USD
10092-214
Anti-PHYH Rabbit Polyclonal Antibody
Antibodies
PHYH (Phytanoyl-CoA 2-hydroxylase), also known as Phytanic acid oxidase or Phytanoyl-CoA alpha-hydroxylase, is a peroxisomal enzyme which converts phytanoyl-CoA to 2-hydroxyphytanoyl-CoA. It catalyzes the first step in the alpha-oxidation of phytanic acid, a branched-chain fatty acid. Mutations in this gene can cause Refsum disease (RD) and deficient protein activity can cause Zellweger syndrome and rhizomelic chondrodysplasia punctata. This antibody (12858-1-AP) detects two bands: 36kd and 70kd. The 70kd band may probably be the dimeric form of PHYH.

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: PHYH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR