Anti-CMIP Rabbit Polyclonal Antibody

Supplier: Proteintech
12851-1-AP-150UL
10084-902EA 402 USD
10084-902
Anti-CMIP Rabbit Polyclonal Antibody
Antibodies
The gene of CMIP was isolated from one patient of minimal change nephrotic syndrome. The gene encodes two isoforms: CMIP and truncated CMIP. The predicted CMIP and truncated CMIP proteins contain 739 and 679 amino acids and have calculated molecular masses of 85 and 75 kD, respectively. The two isoforms have different distribution and expression pattern.

Western Blot: Raji Cells, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: CMIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR