Anti-FBP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12842-1-AP-150UL
10086-894EA 402 USD
10086-894
Anti-FBP1 Rabbit Polyclonal Antibody
Antibodies
FBP1(Fructose-1,6-bisphosphatase 1) is also named as FBP and belongs to the FBPase class 1 family. It catalyzes the hydrolysis of fructose-1,6 bisphosphate to fructose-6-phosphate and inorganic phosphate. This reaction is an important regulatory site of gluconeogenesis. Defects in FBP1 are the cause of fructose-1,6-bisphosphatase deficiency (FBPD).

Western Blot: Human Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: FBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR