Anti-MAPK6 Rabbit Polyclonal Antibody

12839-1-AP
10089-960EA 349.28 USD
10089-960
Anti-MAPK6 Rabbit Polyclonal Antibody
Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: MAPK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR