Anti-KDM3A Rabbit Polyclonal Antibody

Supplier: Proteintech
12835-1-AP-150UL
10088-218EA 599.76 USD
10088-218
Anti-KDM3A Rabbit Polyclonal Antibody
Antibodies
KDM3A, also name as JMJD1A, JHDM2A, JMJD1, JMJD1A, KIAA0742 and TSGA, belongs to the JHDM2 histone demethylase family. It is a histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. It is involved in spermatogenesis by regulating expression of target genes such as PRM1 and TMP1 which are required for packaging and condensation of sperm chromatin. It is involved in obesity resistance through regulation of metabolic genes such as PPARA and UCP1. (PubMed:16603237), JMJD1A is a useful biomarker for hypoxic tumor cells and a prognostic marker that could be a promising therapeutic target against CRC. This antibody recognize the N-term of KDM3A(JMJD1A).

Western Blot: HeLa Cells, 1:1000-1:10000; IF: HepG2 Cells, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: KDM3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR