Anti-BACE Rabbit Polyclonal Antibody

Supplier: Proteintech
12807-1-AP-150UL
10083-474EA 459.7 USD
10083-474
Anti-BACE Rabbit Polyclonal Antibody
Antibodies
beta-secretase, encoded by BACE1(beta-stie amyloid beta A4 precursor protein-cleavage enzyem1) gene, participate in the two step proteolytic processing of the APP. Cleaves at the N-terminus of the A-beta peptide sequence, between residues 671 and 672 of APP, leads to the generation and extracellular release of beta-cleaved soluble APP, and a corresponding cell-associated C-terminal fragment which is later released by gamma-secretase. It has a high expression in the brain and pancreas. This protein has 6 isoforms produced by alternative splicing. BACE1 can exsit as a homodimer with the MW of 120 kDa and the glycosylated immature with the MW of 75 kDa..

  • Western Blot: Mouse Brain Tissue, 1:200-1:2000; IP: Mouse Brain Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: BACE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR