Anti-DNAJA4 Rabbit Polyclonal Antibody

12806-1-AP
10085-930EA 356.38 USD
10085-930
Anti-DNAJA4 Rabbit Polyclonal Antibody
Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: DNAJA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR