Anti-VPS26A Rabbit Polyclonal Antibody

Supplier: Proteintech
12804-1-AP-150UL
10097-012EA 402 USD
10097-012
Anti-VPS26A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:200-1:1000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: VPS26A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR