Anti-GAGE1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12795-1-AP-150UL
10087-314EA 402 USD
10087-314
Anti-GAGE1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: GAGE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR