Anti-CP110 Rabbit Polyclonal Antibody

Supplier: Proteintech
12780-1-AP-150UL
10085-088EA 537.7 USD
10085-088
Anti-CP110 Rabbit Polyclonal Antibody
Antibodies
CP110, also named as CCP110 and KIAA0419, is an 110kDa protein. This gene is different to the gene CEP110 (geneID:11064; CNTRL). CP110 is a centriolar protein that positively regulates centrioleduplication while restricting centriole elongation and ciliogenesis. And it acts as a key negative regulator of ciliogenesis in collaboration with CEP97 by capping the mother centriole thereby preventing cilia formation.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: MEF/HeLa Cells, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000; FC: HeLa Cells, N/A

Type: Primary
Antigen: CP110
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR