Anti-MAFF Rabbit Polyclonal Antibody

Supplier: Proteintech
12771-1-AP-150UL
10089-872EA 537.7 USD
10089-872
Anti-MAFF Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human lung Tissue, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: MAFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR